Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylmorpholine Hydrochloride, 99+%, Thermo Scientific™

Catalog No. AC135112500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC135112500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC135112500 Supplier Thermo Scientific Chemicals Supplier No. 135112500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3651-67-0
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.61
MDL Number MFCD00039042
InChI Key BTJRKNUKPQBLAL-UHFFFAOYSA-N
Synonym n-methylmorpholine hydrochloride, 4-methylmorpholine hydrochloride, 4-methylmorpholine hcl, methylmorpholinium chloride, n-methyl morpholine hydrochloride, n-methylmorpholine hcl, 4-methylmorpholinium chloride, morpholine, 4-methyl-, hydrochloride
PubChem CID 3083815
IUPAC Name hydrogen 4-methylmorpholine chloride
SMILES [H+].[Cl-].CN1CCOCC1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 203°C to 211°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 250 g
Merck Index 10, 6137
Formula Weight 137.61
Percent Purity 99.0 to 101.0%
Physical Form Hygroscopic Powder
Chemical Name or Material N-Methylmorpholine hydrochloride, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Flammable solid.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.