CAS RN 3652-90-2
CAS RN 3652-90-2
IUPAC Name:
2-bromo-9H-carbazole
Synonyms:
2-bromo-9H-carbazole
Molecular Weight (g/mol):
246.11
Molecular Formula:
C12H8BrN
InChi Key:
PJRGCJBBXGNEGD-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)C1=C(N2)C=CC=C1
1
–
1
of
1
results
1
2-Bromocarbazole 98.0+%, TCI America™
CAS: 3652-90-2 Molecular Formula: C12H8BrN Molecular Weight (g/mol): 246.11 MDL Number: MFCD09750430 InChI Key: PJRGCJBBXGNEGD-UHFFFAOYSA-N PubChem CID: 12717089 IUPAC Name: 2-bromo-9H-carbazole SMILES: BrC1=CC2=C(C=C1)C1=C(N2)C=CC=C1