Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromocarbazole 98.0+%, TCI America™
SDP

Catalog No. B43815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43815G 5 g
B438125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43815G Supplier TCI America Supplier No. B43815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3652-90-2
Molecular Formula C12H8BrN
Molecular Weight (g/mol) 246.11
MDL Number MFCD09750430
InChI Key PJRGCJBBXGNEGD-UHFFFAOYSA-N
PubChem CID 12717089
IUPAC Name 2-bromo-9H-carbazole
SMILES BrC1=CC2=C(C=C1)C1=C(N2)C=CC=C1

Specifications

Melting Point 255°C
Color White-Yellow
Quantity 5 g
Formula Weight 246.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromocarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.