accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)OHNOHOCH3CH3OH2O

CAS RN 36546-50-6

IUPAC Name: ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate hydrate
Synonyms: acetyl-L-tyrosine ethyl ester hydrate
Molecular Weight (g/mol): 269.3
Molecular Formula: C13H19NO5
InChi Key: YWAVLHZJMWEYTA-HVPILOLGNA-N
SMILES: O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)