Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-tyrosine ethyl ester monohydrate, 99%

Catalog No. AC102750500
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102750500 50 g Glass Bottle
AC102750010 1 g Glass Bottle
AC102750100 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102750500 Supplier Thermo Scientific Chemicals Supplier No. 102750500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 36546-50-6 | C13H19NO5 | 269.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36546-50-6
Molecular Formula C13H19NO5
Molecular Weight (g/mol) 269.30
MDL Number MFCD00149093
InChI Key YWAVLHZJMWEYTA-HVPILOLGNA-N
Synonym n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
PubChem CID 2723594
IUPAC Name ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
SMILES O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 80°C to 81°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula 4-(HO)C6H4CH2CH(NHCOCH3)CO2C2H5·H2O
Quantity 50 g
Specific Rotation Condition +23° (24°C c=1,C2H5OH)
Specific Rotation + 23
Solubility Information Solubility in water: soluble in cold water. Other solubilities: soluble in hot water,soluble in ethanol
Formula Weight 269.29
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-tyrosine ethyl ester monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.