accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFFFFFFFFFFFFFOFFFFFFFFFFFFFFF

CAS RN 36554-97-9

IUPAC Name: 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,11,11,12,12,13,13,14,14,15,15,16,16,17,17,17-triacontafluoroheptadecane-8,10-dione
Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,11,11,12,12,13,13,14,14,15,15,16,16,17,17,17-triacontafluoroheptadecane-8,10-dione
Molecular Weight (g/mol): 808.15
Molecular Formula: C17H2F30O2
InChi Key: ZXHJIRSEEIEAQG-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(=O)CC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results