Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9H,9H-Triacontafluoro-8,10-heptadecanedione 98.0+%, TCI America™
SDP

Catalog No. T2037100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2037100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2037100MG Supplier TCI America Supplier No. T2037100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36554-97-9
Molecular Formula C17H2F30O2
Molecular Weight (g/mol) 808.153
InChI Key ZXHJIRSEEIEAQG-UHFFFAOYSA-N
Synonym 9H,9H-Perfluoro-8,10-heptadecanedione
PubChem CID 4983346
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,11,11,12,12,13,13,14,14,15,15,16,16,17,17,17-triacontafluoroheptadecane-8,10-dione
SMILES C(C(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)C(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 51°C
Quantity 100 mg
Formula Weight 808.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9H,9H-Triacontafluoro-8,10-heptadecanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.