accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 36556-06-6

IUPAC Name: 5,6,7,8-tetrahydroisoquinoline
Synonyms: 5,6,7,8-tetrahydroisoquinoline
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: HTMGQIXFZMZZKD-UHFFFAOYSA-N
SMILES: C1CCC2=CN=CC=C2C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results