Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6,7,8-Tetrahydroisoquinoline 98.0+%, TCI America™
SDP

Catalog No. T123025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T123025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T123025ML Supplier TCI America Supplier No. T123025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36556-06-6
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.194
MDL Number MFCD00012168
InChI Key HTMGQIXFZMZZKD-UHFFFAOYSA-N
Synonym 5,6,7,8-tetrahydro-isoquinoline, isoquinoline, 5,6,7,8-tetrahydro, acmc-1ct7z, 5,6,7,8-tetra-hydro-isoquinoline, 5,6,7,8-tetrahydroisoquinoline
PubChem CID 119010
IUPAC Name 5,6,7,8-tetrahydroisoquinoline
SMILES C1CCC2=C(C1)C=CN=C2

Specifications

Color Yellow
Boiling Point 81°C
Quantity 25 mL
Formula Weight 133.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5,6,7,8-Tetrahydroisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.