accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3658-77-3

IUPAC Name: 4-hydroxy-2,5-dimethyl-2,3-dihydrofuran-3-one
Synonyms: furaneol dimethylhydroxy furanone
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: INAXVXBDKKUCGI-UHFFFAOYNA-N
SMILES: CC1OC(C)=C(O)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)