Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-4-hydroxy-3(2H)-furanone (15% in Propylene Glycol, ca. 1.2mol/L), TCI America™
SDP

Catalog No. D188425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D188425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D188425ML Supplier TCI America Supplier No. D188425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3658-77-3
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.127
MDL Number MFCD00010706
InChI Key INAXVXBDKKUCGI-UHFFFAOYSA-N
Synonym 4-Hydroxy-2,5-dimethyl-3(2H)-furanone
PubChem CID 19309
ChEBI CHEBI:76247
IUPAC Name 4-hydroxy-2,5-dimethylfuran-3-one
SMILES CC1C(=O)C(=C(O1)C)O

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 128.13
Physical Form Liquid
Chemical Name or Material 2,5-Dimethyl-4-hydroxy-3(2H)-furanone (15% in Propylene Glycol, ca. 1.2mol/L)
EINECSNumber (5)-5509
RTECSNumber LU3990000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.