accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3658-93-3

IUPAC Name: 1,1-diethoxyhexane
Synonyms: hexanal diethyl acetal
Molecular Weight (g/mol): 174.28
Molecular Formula: C10H22O2
InChi Key: WNHOMUCDFNTSEV-UHFFFAOYSA-N
SMILES: CCCCCC(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results