accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3COCH3

CAS RN 3658-95-5

IUPAC Name: 1,1-diethoxybutane
Synonyms: 1,1-diethoxybutane
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: UVHXZFGCCJLFMX-UHFFFAOYSA-N
SMILES: CCCC(OCC)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)