accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOO

CAS RN 3669-80-5

IUPAC Name: 11-hydroxyundecanoate
Synonyms: 11-hydroxyundecanoate
Molecular Weight (g/mol): 201.29
Molecular Formula: C11H21O3
InChi Key: KNRCBASNXNXUQQ-UHFFFAOYSA-M
SMILES: OCCCCCCCCCCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results