accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36703-88-5

IUPAC Name: tris(1-(dimethylamino)propan-2-ol); tris(4-acetamidobenzoic acid); 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms: inosine pranobex
Molecular Weight (g/mol): 1115.25
Molecular Formula: C52H78N10O17
InChi Key: YLDCUKJMEKGGFI-QCSRICIXSA-N
SMILES: CC(O)CN(C)C.CC(O)CN(C)C.CC(O)CN(C)C.CC(=O)NC1=CC=C(C=C1)C(O)=O.CC(=O)NC1=CC=C(C=C1)C(O)=O.CC(=O)NC1=CC=C(C=C1)C(O)=O.OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1NC=NC2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)