accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ONOHOOHOHNNNH2O

CAS RN 36707-00-3

IUPAC Name: 4-amino-8-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-oxo-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxamide
Synonyms: 4-amino-8-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-oxopyrido[2,3-d]pyrimidine-6-carboxamide
Molecular Weight (g/mol): 337.29
Molecular Formula: C13H15N5O6
InChi Key: SPBWHPXCWJLQRU-UHFFFAOYNA-N
SMILES: NC(=O)C1=CN(C2OC(CO)C(O)C2O)C2=NC=NC(N)=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results