Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

API-1, R&D Systems™
SDP

Catalog No. 389710 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-971-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 38-971-0 Supplier R&D Systems Supplier No. 3897/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective Akt/PKB inhibitor. Antitumor

Akt/protein kinase B (PKB) inhibitor. Binds the pleckstrin homology domain of Akt and blocks Akt membrane translocation. Inhibits EGF-induced kinase activity of Akt1, Akt2 and Akt3. Induces cell growth arrest and apoptosis in human cancer cells expressing constitutively active Akt. Displays antitumor activity in vitro and in vivo.

Chemical Identifiers

CAS 36707-00-3
Molecular Formula C13H15N5O6
Molecular Weight (g/mol) 337.292
InChI Key SPBWHPXCWJLQRU-FITJORAGSA-N
Synonym api-1, 4-amino-8-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-oxo, 4-amino-5,8-dihydro-5-oxo-8-?-d-ribofuranosyl-pyrido 2,3-d pyrimidine-6-carboxamide, 4-amino-5,8-dihydro-5-oxo-8-beta-d-ribofuranosyl-pyrido 2,3-d pyrimidine-6-carboxamide, 4-amino-8-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-oxopyrido 2,3-d pyrimidine-6-carboxamide
PubChem CID 24773090
IUPAC Name 4-amino-8-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-oxopyrido[2,3-d]pyrimidine-6-carboxamide
SMILES C1=C(C(=O)C2=C(N1C3C(C(C(O3)CO)O)O)N=CN=C2N)C(=O)N

Specifications

Quantity 10 mg
Formula Weight 337.29
Percent Purity >96%
Product Line Tocris
Chemical Name or Material API-1
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.