CAS RN 3674-18-8
CAS RN 3674-18-8
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
Synonyms:
2,5-dioxopyrrolidin-1-yl 2-[(tert-butoxycarbonyl)amino]-3-phenylpropanoate
Molecular Weight (g/mol):
362.38
Molecular Formula:
C18H22N2O6
InChi Key:
NHUCANAMPJGMQL-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Phe-OSu, Advanced ChemTech
CAS: 3674-18-8 Molecular Formula: C18H22N2O6 Molecular Weight (g/mol): 362.38 InChI Key: NHUCANAMPJGMQL-UHFFFAOYNA-N IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O