CAS RN 36775-23-2
IUPAC Name:
2-chloro-N-(1-hydroxy-2,2,6,6-tetramethylpiperidin-4-yl)acetamide
Synonyms:
2-chloro-N-(1-hydroxy-2,2,6,6-tetramethylpiperidin-4-yl)acetamide
Molecular Weight (g/mol):
248.75
Molecular Formula:
C11H21ClN2O2
InChi Key:
ICXMAAHRVIPICX-UHFFFAOYSA-N
SMILES:
CC1(C)CC(CC(C)(C)N1O)NC(=O)CCl
1
–
1
of
1
results
1
4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 96.0+%, TCI America™
CAS: 36775-23-2 Molecular Formula: C11H20ClN2O2 MDL Number: MFCD00041858 Synonym: 4-(2-Chloroacetamido)-TEMPO