Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 96.0+%, TCI America™
SDP

Catalog No. C14321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14321G 1 g
C1432100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14321G Supplier TCI America Supplier No. C14321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36775-23-2
Molecular Formula C11H20ClN2O2
MDL Number MFCD00041858
Synonym 4-(2-Chloroacetamido)-TEMPO

Specifications

Melting Point 117°C
Quantity 1 g
Formula Weight 247.74
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.