CAS RN 3679-64-9
IUPAC Name:
5-bromo-N-(4-chlorophenyl)-2-hydroxybenzamide
Synonyms:
bromochlorosalicylanilide 4'-CL-5-BR salicylanilide
Molecular Weight (g/mol):
326.57
Molecular Formula:
C13H9BrClNO2
InChi Key:
QBSGXIBYUQJHMJ-UHFFFAOYSA-N
SMILES:
OC1=CC=C(Br)C=C1C(=O)NC1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
5-Bromo-4'-chlorosalicylanilide 98.0+%, TCI America™
CAS: 3679-64-9 Molecular Formula: C13H9BrClNO2 Molecular Weight (g/mol): 326.57 MDL Number: MFCD00020132 InChI Key: QBSGXIBYUQJHMJ-UHFFFAOYSA-N Synonym: 5-Bromo-4′C-chloro-2-hydroxybenzanilide PubChem CID: 77254 IUPAC Name: 5-bromo-N-(4-chlorophenyl)-2-hydroxybenzamide SMILES: OC1=CC=C(Br)C=C1C(=O)NC1=CC=C(Cl)C=C1