Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4'-chlorosalicylanilide 98.0+%, TCI America™
SDP

Catalog No. B089710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B089710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B089710G Supplier TCI America Supplier No. B089710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3679-64-9
Molecular Formula C13H9BrClNO2
Molecular Weight (g/mol) 326.57
MDL Number MFCD00020132
InChI Key QBSGXIBYUQJHMJ-UHFFFAOYSA-N
Synonym 5-Bromo-4′C-chloro-2-hydroxybenzanilide
PubChem CID 77254
IUPAC Name 5-bromo-N-(4-chlorophenyl)-2-hydroxybenzamide
SMILES OC1=CC=C(Br)C=C1C(=O)NC1=CC=C(Cl)C=C1

Specifications

Melting Point 246°C
Color White-Yellow
Quantity 10 g
Formula Weight 326.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4′-chlorosalicylanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.