accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 368-43-4

IUPAC Name: benzenesulfonyl fluoride
Synonyms: benzenesulfonyl fluoride
Molecular Weight (g/mol): 160.16
Molecular Formula: C6H5FO2S
InChi Key: IDIPWEYIBKUDNY-UHFFFAOYSA-N
SMILES: FS(=O)(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)