accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3680-69-1

ClNNHN

CAS RN 3680-69-1

IUPAC Name: 4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Synonyms: 4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol): 153.57
Molecular Formula: C6H4ClN3
InChi Key: BPTCCCTWWAUJRK-UHFFFAOYSA-N
SMILES: ClC1=C2C=CNC2=NC=N1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (4)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (4)