CAS RN 3682-35-7
2,4,6-Tripyridyl-S-triazine, 101.0%, MP Biomedicals™
CAS: 3682-35-7 Molecular Formula: C18H12N6 Molecular Weight (g/mol): 312.34 MDL Number: MFCD00006045 InChI Key: KMVWNDHKTPHDMT-UHFFFAOYSA-N Synonym: 2,4,6-tripyridyl-s-triazine,2,4,6-tris 2-pyridyl-s-triazine,2,4,6-tri 2-pyridyl-1,3,5-triazine,terpyridyl-s-triazine,2,4,6-tri 2-pyridyl-s-triazine,tptz iron reagent,1,3,5-triazine, 2,4,6-tri-2-pyridinyl,tri-2-pyridyl-s-triazine,s-triazine, tri-2-pyridyl,2,4,6-tris 2-pyridyl-1,3,5-triazine PubChem CID: 77258 IUPAC Name: tris(pyridin-2-yl)-1,3,5-triazine SMILES: C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
2,4,6-Tris(2-pyridyl)-s-triazine, For Spectrophotometric Det. of Fe, 99.0% (HPLC), MilliporeSigma™ Supelco™
CAS: 3682-35-7 Molecular Formula: C18H12N6 Molecular Weight (g/mol): 312.34 MDL Number: MFCD00006045 InChI Key: KMVWNDHKTPHDMT-UHFFFAOYSA-N Synonym: 2,4,6-Tri(2-pyridyl)-s-triazine; TPTZ IUPAC Name: tris(pyridin-2-yl)-1,3,5-triazine SMILES: C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
2,4,6-Tris(2-pyridyl)-s-triazine, For Spectrophotometric Det. of Fe, 98%, MilliporeSigma™ Supelco™
CAS: 3682-35-7 Molecular Formula: C18H12N6 Molecular Weight (g/mol): 312.34 MDL Number: MFCD00006045 InChI Key: KMVWNDHKTPHDMT-UHFFFAOYSA-N Synonym: 2,4,6-Tri(2-pyridyl)-s-triazine; TPTZ IUPAC Name: tris(pyridin-2-yl)-1,3,5-triazine SMILES: C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
2,4,6-Tri(2-pyridyl)-s-triazine, 99%
CAS: 3682-35-7 Molecular Formula: C18H12N6 Molecular Weight (g/mol): 312.34 MDL Number: MFCD00006045 InChI Key: KMVWNDHKTPHDMT-UHFFFAOYSA-N Synonym: 2,4,6-tripyridyl-s-triazine,2,4,6-tris 2-pyridyl-s-triazine,2,4,6-tri 2-pyridyl-1,3,5-triazine,terpyridyl-s-triazine,2,4,6-tri 2-pyridyl-s-triazine,tptz iron reagent,1,3,5-triazine, 2,4,6-tri-2-pyridinyl,tri-2-pyridyl-s-triazine,s-triazine, tri-2-pyridyl,2,4,6-tris 2-pyridyl-1,3,5-triazine PubChem CID: 77258 SMILES: C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%
CAS: 3682-35-7 Molecular Formula: C18H12N6 Molecular Weight (g/mol): 312.34 MDL Number: MFCD00006045 InChI Key: KMVWNDHKTPHDMT-UHFFFAOYSA-N Synonym: 2,4,6-tripyridyl-s-triazine,2,4,6-tris 2-pyridyl-s-triazine,2,4,6-tri 2-pyridyl-1,3,5-triazine,terpyridyl-s-triazine,2,4,6-tri 2-pyridyl-s-triazine,tptz iron reagent,1,3,5-triazine, 2,4,6-tri-2-pyridinyl,tri-2-pyridyl-s-triazine,s-triazine, tri-2-pyridyl,2,4,6-tris 2-pyridyl-1,3,5-triazine PubChem CID: 77258 SMILES: C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
2,4,6-Tri(2-pyridyl)-1,3,5-triazine 98.0+%, TCI America™
CAS: 3682-35-7 Molecular Formula: C18H12N6 Molecular Weight (g/mol): 312.34 MDL Number: MFCD00006045 InChI Key: KMVWNDHKTPHDMT-UHFFFAOYSA-N Synonym: 2,4,6-tripyridyl-s-triazine,2,4,6-tris 2-pyridyl-s-triazine,2,4,6-tri 2-pyridyl-1,3,5-triazine,terpyridyl-s-triazine,2,4,6-tri 2-pyridyl-s-triazine,tptz iron reagent,1,3,5-triazine, 2,4,6-tri-2-pyridinyl,tri-2-pyridyl-s-triazine,s-triazine, tri-2-pyridyl,2,4,6-tris 2-pyridyl-1,3,5-triazine PubChem CID: 77258 IUPAC Name: tris(pyridin-2-yl)-1,3,5-triazine SMILES: C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1