Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tris(2-pyridyl)-s-triazine, For Spectrophotometric Det. of Fe, 98%, MilliporeSigma™ Supelco™

Catalog No. 111014057 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-4057 5 g Glass Bottle
11-101-4054 1 g Glass Bottle
11-101-4056 10 g Glass Bottle
11-101-4055 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-101-4057 Supplier MilliporeSigma Supelco Supplier No. T12535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4,6-Tris(2-pyridyl)-s-triazine is a complexing reagent used in spectrophotometric analysis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3682-35-7
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.34
MDL Number MFCD00006045
InChI Key KMVWNDHKTPHDMT-UHFFFAOYSA-N
Synonym 2,4,6-Tri(2-pyridyl)-s-triazine; TPTZ
IUPAC Name tris(pyridin-2-yl)-1,3,5-triazine
SMILES C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 247°C to 249°C
Packaging Glass Bottle
Linear Formula C18H12N6
Quantity 5 g
Formula Weight 312.33
Percent Purity ≥98%
Grade For spectrophotometric det. (of Fe)
Physical Form Crystalline Powder
RTECSNumber XZ2050000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.