accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHNSOOONHNSOOOOSOOONaNaNa(E/Z)(E/Z)

CAS RN 3682-47-1

IUPAC Name: trisodium 4-oxo-5-[2-(2-oxo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]-3-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: fast sulphon black F
Molecular Weight (g/mol): 774.63
Molecular Formula: C30H17N4Na3O11S3
InChi Key: MKMBAZXKTDYGAC-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC(NN=C2C(=O)C=CC3=CC=CC=C23)=C2C(=O)C(=NNC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=CC2=C1)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results