Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Sulphon Black F, Thermo Scientific™

Catalog No. AC339350050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC339350050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC339350050 Supplier Thermo Scientific Chemicals Supplier No. 339350050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3682-47-1
Molecular Formula C30H17N4Na3O11S3
Molecular Weight (g/mol) 774.632
MDL Number MFCD00021612
InChI Key MKMBAZXKTDYGAC-LQSSYBFTSA-K
Synonym Acid Black 32, C.I. 26990
PubChem CID 50987236
IUPAC Name trisodium;(3E)-4-oxo-5-[(2E)-2-(2-oxonaphthalen-1-ylidene)hydrazinyl]-3-[(4-sulfonatonaphthalen-1-yl)hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=C4C(=CC(=C3)S(=O)(=O)[O-])C=C(C(=NNC5=CC=C(C6=CC=CC=C65)S(=O)(=O)[O-])C4=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Infrared Spectrum Authentic
Formula Weight 774.63
Color Black
Quantity 5 g
Physical Form Powder
Chemical Name or Material Fast Sulphon Black F
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.