CAS RN 368449-04-1
CAS RN 368449-04-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Telomerase Inhibitor IX
The Telomerase Inhibitor IX, also referenced under CAS 368449-04-1, controls the biological activity of Telomerase. This small molecule/inhibitor is primarily used for Cell Structure applications.