accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBr

CAS RN 36865-41-5

IUPAC Name: 1-bromo-3-methoxypropane
Synonyms: 1-bromo-3-methoxypropane
Molecular Weight (g/mol): 153.02
Molecular Formula: C4H9BrO
InChi Key: CEVMYGZHEJSOHZ-UHFFFAOYSA-N
SMILES: COCCCBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results