CAS RN 36875-26-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Trifluoroacetyl-D-glucosamine 96.0+%, TCI America™
CAS: 36875-26-0 Molecular Formula: C8H12F3NO6 MDL Number: MFCD00059807 Synonym: 2-Deoxy-2-trifluoroacetamido-D-glucopyranose