Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Trifluoroacetyl-D-glucosamine 96.0+%, TCI America™
SDP

Catalog No. T09735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09735G 5 g
T09731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09735G Supplier TCI America Supplier No. T09735G

Chemical Identifiers

CAS 36875-26-0
Molecular Formula C8H12F3NO6
MDL Number MFCD00059807
Synonym 2-Deoxy-2-trifluoroacetamido-D-glucopyranose

Specifications

Quantity 5 g
Formula Weight 275.18
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Trifluoroacetyl-D-glucosamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.