accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2NNOO

CAS RN 3694-52-8

IUPAC Name: 3-nitrobenzene-1,2-diamine
Synonyms: 3-nitro-O-phenylenediamine
Molecular Weight (g/mol): 153.14
Molecular Formula: C6H7N3O2
InChi Key: IOCXBXZBNOYTLQ-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1N)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)