accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3695-24-7

IUPAC Name: (2S)-2-hydroxy-2-(3-hydroxy-4-methoxyphenyl)acetate
Synonyms: (2S)-2-hydroxy-2-(3-hydroxy-4-methoxyphenyl)acetate
Molecular Weight (g/mol): 197.17
Molecular Formula: C9H9O5
InChi Key: PXMUSCHKJYFZFD-QMMMGPOBSA-M
SMILES: COC1=CC=C(C=C1O)[C@H](O)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results