Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3-Hydroxy-4-methoxymandelic Acid 99%, Thermo Scientific™

Catalog No. AC216262500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC216262500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC216262500 Supplier Thermo Scientific Chemicals Supplier No. 216262500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3695-24-7
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
InChI Key PXMUSCHKJYFZFD-UHFFFAOYSA-N
Synonym 3-hydroxy-4-methoxymandelic acid, 2-hydroxy-2-3-hydroxy-4-methoxyphenyl acetic acid, hydroxy 3-hydroxy-4-methoxyphenyl acetic acid, dl-3-hydroxy-4-methoxymandelic acid, 2s-2-hydroxy-2-3-hydroxy-4-methoxy-phenyl acetate, 2-3'-hydroxy-4'-methoxyphenyl-2-hydroxyethanoic acid, 3-hydroxy-4-methoxymandelicacid, 3-hydroxy-4-methoxy-mandelic acid, hydroxy 3-hydroxy-4-methoxyphenyl acetic acid #
PubChem CID 107153
IUPAC Name 2-hydroxy-2-(3-hydroxy-4-methoxyphenyl)acetic acid
SMILES COC1=C(C=C(C=C1)C(C(=O)O)O)O

Specifications

Melting Point 131°C to 134°C
Color White to Beige
Infrared Spectrum Authentic
Linear Formula HOC6H3(OCH3)CH(OH)CO2H
Quantity 250 mg
Beilstein 10, III, 2100
Formula Weight 198.17
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-3-Hydroxy-4-methoxymandelic acid, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.