accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3H3CNHOHOHNHSCH3OO

CAS RN 37000-20-7

IUPAC Name: hydrogen N-(2-hydroxy-5-{1-hydroxy-2-[(2-methyl-1-phenylpropan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride
Synonyms: zinterol
Molecular Weight (g/mol): 414.95
Molecular Formula: C19H27ClN2O4S
InChi Key: LVNUBJDWJFOMKH-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)(CC1=CC=CC=C1)NCC(O)C1=CC=C(O)C(NS(C)(=O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results