accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHN

CAS RN 37059-57-7

IUPAC Name: N-ethylpyridin-2-amine
Synonyms: N-ethylpyridin-2-amine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: CXGFWBPQQXZELI-UHFFFAOYSA-N
SMILES: CCNC1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results