Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Ethylamino)pyridine 98.0+%, TCI America™
SDP

Catalog No. E05585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05585G 5 g
E055825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05585G Supplier TCI America Supplier No. E05585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 37059-57-7 | C7H10N2 | 122.171 g/mol

Chemical Identifiers

CAS 37059-57-7
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD04038418
InChI Key CXGFWBPQQXZELI-UHFFFAOYSA-N
PubChem CID 11804745
IUPAC Name N-ethylpyridin-2-amine
SMILES CCNC1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Ethylamino)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.