accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3

CAS RN 37064-20-3

IUPAC Name: propyl 2-methylbutanoate
Synonyms: propyl 2-methylbutanoate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: TZFQMSDUSOTCJC-UHFFFAOYNA-N
SMILES: CCCOC(=O)C(C)CC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results