Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl 2-Methylbutyrate 98.0+%, TCI America™
SDP

Catalog No. P180925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P180925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P180925ML Supplier TCI America Supplier No. P180925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37064-20-3
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00085211
InChI Key TZFQMSDUSOTCJC-UHFFFAOYSA-N
Synonym 2-Methylbutyric Acid Propyl Ester
PubChem CID 162239
IUPAC Name propyl 2-methylbutanoate
SMILES CCCOC(=O)C(C)CC

Specifications

Color Colorless
Boiling Point 157°C
Quantity 25 mL
UN Number 3272
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl 2-Methylbutyrate
EINECSNumber (2)-0776
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.