accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOHNCH3

CAS RN 3708-39-2

IUPAC Name: N-methyl-4-[4-(methylamino)benzoyl]aniline
Synonyms: N-methyl-4-[4-(methylamino)benzoyl]aniline
Molecular Weight (g/mol): 240.31
Molecular Formula: C15H16N2O
InChi Key: HXTBYXIZCDULQI-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1)C(=O)C1=CC=C(NC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results