Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(methylamino)benzophenone 98.0+%, TCI America™
SDP

Catalog No. B14331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14331G Supplier TCI America Supplier No. B14331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3708-39-2
Molecular Formula C15H16N2O
Molecular Weight (g/mol) 240.306
MDL Number MFCD00143227
InChI Key HXTBYXIZCDULQI-UHFFFAOYSA-N
PubChem CID 584435
IUPAC Name bis[4-(methylamino)phenyl]methanone
SMILES CNC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)NC

Specifications

Melting Point 133°C
Color Yellow
Quantity 1 g
Formula Weight 240.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(methylamino)benzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.