accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 370865-80-8

IUPAC Name: 1-butyl-1-methylpyrrolidin-1-ium; [(cyanoimino)methylidene]azanide
Synonyms: 1-butyl-1-methylpyrrolidin-1-ium; [(cyanoimino)methylidene]azanide
Molecular Weight (g/mol): 208.31
Molecular Formula: C11H20N4
InChi Key: LBHLGZNUPKUZJC-UHFFFAOYSA-N
SMILES: [N-]=C=NC#N.CCCC[N+]1(C)CCCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results