Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-1-methylpyrrolidinium Dicyanamide 98.0+%, TCI America™
SDP

Catalog No. B54535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54535G 5 g
B545325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54535G Supplier TCI America Supplier No. B54535G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 370865-80-8
Molecular Formula C11H20N4
Molecular Weight (g/mol) 208.309
MDL Number MFCD08458923
InChI Key LBHLGZNUPKUZJC-UHFFFAOYSA-N
PubChem CID 16213711
IUPAC Name 1-butyl-1-methylpyrrolidin-1-ium;cyanoiminomethylideneazanide
SMILES CCCC[N+]1(CCCC1)C.C(=[N-])=NC#N

Specifications

Color Yellow
Quantity 5 g
Formula Weight 208.31
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-1-methylpyrrolidinium Dicyanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.