accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOFFF

CAS RN 371-27-7

IUPAC Name: 2,2,2-trifluoroethyl butanoate
Synonyms: 2,2,2-trifluoroethyl butanoate
Molecular Weight (g/mol): 170.13
Molecular Formula: C6H9F3O2
InChi Key: DEXWRCYOMLUJRF-UHFFFAOYSA-N
SMILES: CCCC(=O)OCC(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results