accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3710-31-4

IUPAC Name: heptane-1,2-diol
Synonyms: heptane-1,2-diol
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: GCXZDAKFJKCPGK-UHFFFAOYNA-N
SMILES: CCCCCC(O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results