Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Heptanediol 98.0+%, TCI America™
SDP

Catalog No. H09485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09485G Supplier TCI America Supplier No. H09485G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3710-31-4
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.20
MDL Number MFCD01861287
InChI Key GCXZDAKFJKCPGK-UHFFFAOYNA-N
Synonym 1,2-Dihydroxyheptane
PubChem CID 77302
IUPAC Name heptane-1,2-diol
SMILES CCCCCC(O)CO

Specifications

Color Colorless
Boiling Point 130°C
Quantity 5 g
Formula Weight 132.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Heptanediol
EINECSNumber (2)-0240
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.