accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNOHNOH(Z)(Z)

CAS RN 37110-24-0

IUPAC Name: N-[(1Z,5Z)-5-(hydroxyimino)-3,3-dimethylcyclohexylidene]hydroxylamine
Synonyms: N-[(1Z,5Z)-5-(hydroxyimino)-3,3-dimethylcyclohexylidene]hydroxylamine
Molecular Weight (g/mol): 170.21
Molecular Formula: C8H14N2O2
InChi Key: INCDZDUOXAORNG-KZZDLZNXSA-N
SMILES: CC1(C)C\C(C\C(C1)=N/O)=N\O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results