Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimedone Dioxime [for Determination of Co], TCI America™
SDP

Catalog No. D14001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D14001G Supplier TCI America Supplier No. D14001G

Chemical Identifiers

CAS 37110-24-0
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.21
MDL Number MFCD00059877
InChI Key INCDZDUOXAORNG-KZZDLZNXSA-N
Synonym 5,5-Dimethyl-1,3-cyclohexanedione Dioxime, Dimethyldihydroresorcinol Dioxime, Methone Dioxime
PubChem CID 9562213
IUPAC Name N-[(1Z,5Z)-5-(hydroxyimino)-3,3-dimethylcyclohexylidene]hydroxylamine
SMILES CC1(C)C\C(C\C(C1)=N/O)=N\O

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 1 g
Formula Weight 170.21
Physical Form Crystalline Powder
Chemical Name or Material Dimedone Dioxime [for Determination of Co]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.