CAS RN 371242-69-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ PI 3-Kδ Inhibitor X, IC87114
The PI 3-Kδ Inhibitor X, IC87114, also referenced under CAS 371242-69-2, controls the biological activity of PI 3-Kδ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
2
Globe Scientific Cap Insert
Cap inserts suitable for use with RingSeal Cryogenic Vials